Ingenol 99+%

Supplier: Ambeed
Danger

Synonyms: (1aR,2S,5R,5aR,6S,8aS,9R,10aR)-5,5a,6-Trihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-1a,2,5,5a,6,9,10,10a-octahydro-1H-2,8a-methanocyclopenta[a]cyclopropa[e][10]annulen-11-one

A689957-10MG A689957-1MG A689957-25MG A689957-50MG A689957-5MG A689957-1G
76806-642EA 14.86 USD
76806-642 76806-644 76806-646 76806-648 76806-650 77602-754
Ingenol 99+%
Ingenol

Precaución: for Research Use Only

Formula: C₂₀H₂₈O₅
MW: 348.43 g/mol
MDL Number: MFCD27980538
CAS Number: 30220-46-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR