β-Ionone 95%

Supplier: Ambeed
Danger

Synonyms: 4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, 4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one

A686767-25G A686767-500G A686767-1000G
76806-628EA 9.55 USD
76806-628 76806-630 77669-870
β-Ionone 95%
β-Ionone

Precaución: for Research Use Only

Formula: C₁₃H₂₀O
MW: 192.30 g/mol
MDL Number: MFCD00167092
CAS Number: 14901-07-6
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR