Dihydrogen hexahydroxyplatinate(IV) 98%

Supplier: Ambeed
Warning

A681956-100MG A681956-1G A681956-250MG
76806-604EA 20 USD
76806-604 76806-606 76806-608
Dihydrogen hexahydroxyplatinate(IV) 98%
Dihydrogen hexahydroxyplatinate(IV)

Precaución: for Research Use Only

Formula: H₂Pt(OH)₆
MDL Number: MFCD00058744
CAS Number: 51850-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR