Hydrazinium monochloride 97%

Supplier: Ambeed
Danger

Synonyms: Hydrazinium monochloride, Hydrazinium chloride, Hydrazine monohydrochloride, Hydrazine hydrochloride

A681687-10G A681687-25G
76806-600EA 6 USD
76806-600 76806-602
Hydrazinium monochloride 97%
Hydrazinium monochloride

Precaución: for Research Use Only

Formula: H₅ClN₂
MDL Number: MFCD00044368
CAS Number: 2644-70-4
UN: 3290
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR