Bromocresol green indicator

Supplier: Ambeed
Warning

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

A681493-100G A681493-10G A681493-25G A681493-5G A681493-500G
76806-592EA 98.67 USD
76806-592 76806-594 76806-596 76806-598 77602-734
Bromocresol green indicator
Bromocresol green

Precaución: for Research Use Only

Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
MDL Number: MFCD00005874
CAS Number: 76-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR