Ethyl-3-phenylbut-2-enoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl trans-beta-methylcinnamate, Ethyl 3-phenylbut-2-enoate

A681456-1G A681456-250MG
76806-588EA 119.89 USD
76806-588 76806-590
Ethyl-3-phenylbut-2-enoate 98%
Ethyl-3-phenylbut-2-enoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
MDL Number: MFCD00053762
CAS Number: 945-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR