N-Fmoc-DL-pipecolic acid 98%

Supplier: Ambeed
Danger

Synonyms: 1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid

A677789-1G A677789-250MG A677789-5G
76806-582EA 60.48 USD
76806-582 76806-584 76806-586
N-Fmoc-DL-pipecolic acid 98%
N-Fmoc-DL-pipecolic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
MDL Number: MFCD01823265
CAS Number: 105751-19-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR