Lithium tetrafluoroborate 99%

Supplier: Ambeed
Danger

A674457-25G A674457-5G
76806-568EA 30 USD
76806-568 76806-570
Lithium tetrafluoroborate 99%
Lithium tetrafluoroborate

Precaución: for Research Use Only

Formula: BF₄Li
MDL Number: MFCD00011087
CAS Number: 14283-07-9
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR