Methyl-3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

A670112-1G A670112-250MG A670112-25G A670112-5G
76806-528EA 43 USD
76806-528 76806-530 76806-532 76806-534
Methyl-3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate 98%
Methyl-3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO₂S
MDL Number: MFCD00052591
CAS Number: 91076-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR