Margaric acid 98%
Supplier: Ambeed
|
Warning
|
A669114-100G
A669114-10G
A669114-1G
A669114-5G
A669114-25G
76806-518EA
242
USD
76806-518
76806-520
76806-522
76806-524
77880-401
Margaric acid 98%
Margaric acid
Precaución: for Research Use Only
Formula:
C₁₇H₃₄O₂ |
MDL Number:
MFCD00002751 CAS Number: 506-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...