Methyl-11-bromoundecanoate 98%

Supplier: Ambeed
Warning

Synonyms: 11-Bromoundecanoic Acid Methyl Ester, Methyl 11-bromoundecanoate

A664529-100G A664529-1G A664529-250MG A664529-25G A664529-5G
76806-474EA 494.38 USD
76806-474 76806-476 76806-478 76806-480 76806-482
Methyl-11-bromoundecanoate 98%
Methyl-11-bromoundecanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₃BrO₂
MW: 279.22 g/mol
MDL Number: MFCD00045047
CAS Number: 6287-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR