2,6-Dibromo-1,5-dihydroxynaphthalene 93%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dibromonaphthalene-1,5-diol, 2,6-Dibromo-1,5-naphthalenediol

A662347-5G A662347-25G
76806-466EA 151 USD
76806-466 77880-391
2,6-Dibromo-1,5-dihydroxynaphthalene 93%
2,6-Dibromo-1,5-dihydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂O₂
MW: 317.96 g/mol
MDL Number: MFCD00021473
CAS Number: 84-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR