Ethyl-5-methyl[1,2,4]oxadiazole-3-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-methyl-1,2,4-oxadiazole-3-carboxylate

A661081-100MG A661081-1G A661081-250MG A661081-5G
76806-454EA 20.16 USD
76806-454 76806-456 76806-458 76806-460
Ethyl-5-methyl[1,2,4]oxadiazole-3-carboxylate 96%
Ethyl-5-methyl[1,2,4]oxadiazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
MDL Number: MFCD09864941
CAS Number: 40699-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR