Tris(4-formylphenyl)amine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4',4''-Nitrilotribenzaldehyde
A659624-100MG
A659624-1G
A659624-250MG
A659624-5G
76806-446EA
9
USD
76806-446
76806-448
76806-450
76806-452
Tris(4-formylphenyl)amine 95%
Tris(4-formylphenyl)amine
Precaución: for Research Use Only
Formula:
C₂₁H₁₅NO₃ |
MDL Number:
MFCD08276437 CAS Number: 119001-43-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...