Diphenylmethane-4,4'-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A657069-1G A657069-250MG A657069-25G
76806-408EA 36 USD
76806-408 76806-410 77880-379
Diphenylmethane-4,4'-dicarboxylic acid 97%
Diphenylmethane-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₄
MDL Number: MFCD00060322
CAS Number: 790-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR