Fmoc-1-aminocyclobutane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclobutanecarboxylic acid

A654439-1G A654439-250MG A654439-5G A654439-25G
76806-390EA 12 USD
76806-390 76806-392 76806-394 77551-984
Fmoc-1-aminocyclobutane-1-carboxylic acid 98%
Fmoc-1-aminocyclobutane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₄
MDL Number: MFCD02682624
CAS Number: 885951-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR