3-Methyl-8-quinolinesulfonyl chloride 95%

Supplier: Ambeed
Danger

A654064-1G A654064-25G A654064-5G
76806-384EA 10.61 USD
76806-384 76806-386 76806-388
3-Methyl-8-quinolinesulfonyl chloride 95%
3-Methyl-8-quinolinesulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂S
MW: 241.69 g/mol
MDL Number: MFCD02683367
CAS Number: 74863-82-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR