Imidazo[1,5-a]pyridine-7-carboxylic acid 97%

Supplier: Ambeed

A653014-100MG A653014-1G A653014-250MG
76806-378EA 40 USD
76806-378 76806-380 76806-382
Imidazo[1,5-a]pyridine-7-carboxylic acid 97%
Imidazo[1,5-a]pyridine-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD11870763
CAS Number: 588720-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR