Copper(II) disodium ethylenediaminetetraacetate tetrahydrate 98%

Supplier: Ambeed
Warning

Synonyms: Ethylenediaminetetraacetic acid Copper(II) disodium salt, Copper(II) disodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate tetrahydrate, Copper Disodium Ethylenediaminetetraacetate hydrate

A648804-100G A648804-500G
76806-358EA 8 USD
76806-358 76806-360
Copper(II) disodium ethylenediaminetetraacetate tetrahydrate 98%
Copper(II) disodium ethylenediaminetetraacetate tetrahydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₂CuN₂Na₂O₈·4H₂O
MW: 469.80 g/mol
MDL Number: MFCD00148920
CAS Number: 39208-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR