Cresol red ≥93% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: o-Cresolsulphonphthalein, 3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

A642655-100G A642655-25G
76806-324EA 37.14 USD
76806-324 77594-972
Cresol red ≥93% (by HPLC)
Cresol red

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₅S
MW: 382.43 g/mol
MDL Number: MFCD00005878
CAS Number: 1733-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR