3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A641623-1G A641623-5G
76806-314EA 24.41 USD
76806-314 76806-316
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid 98%
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₇ClFNO₃
MW: 255.63 g/mol
MDL Number: MFCD00055659
CAS Number: 3919-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR