L-Methionine sulfoximine 98%

Supplier: Ambeed
Warning

Synonyms: (2S)-2-Amino-4-(S-methylsulphonimidoyl)butanoic acid, H-Met(sulphoximine)-OH

A640782-100MG A640782-1G A640782-250MG A640782-5G
76806-308EA 75.33 USD
76806-308 76806-310 76806-312 77669-738
L-Methionine sulfoximine 98%
L-Methionine sulfoximine

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O₃S
MW: 180.222 g/mol
MDL Number: MFCD00002621
CAS Number: 15985-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR