1,1,1,5,5,5-Hexafluoroacetylacetone 97%

Supplier: Ambeed
Danger

Synonyms: Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA

A637749-10G A637749-1G A637749-25G A637749-5G
76806-280EA 36.08 USD
76806-280 76806-282 77551-938 77602-614
1,1,1,5,5,5-Hexafluoroacetylacetone 97%
1,1,1,5,5,5-Hexafluoroacetylacetone

Precaución: for Research Use Only

Formula: C₅H₂F₆O₂
MW: 208.06 g/mol
MDL Number: MFCD00000426
CAS Number: 1522-22-1
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR