Sodium methotrexate 98%

Supplier: Ambeed
Danger

Synonyms: Sodium (S)-2-(4-(((2,4-diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioate, Methotrexate disodium salt

A637169-25G A637169-5G
76806-268EA 50.93 USD
76806-268 76806-270
Sodium methotrexate 98%
Sodium methotrexate

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₈Na₂O₅
MW: 498.411 g/mol
MDL Number: MFCD00274560
CAS Number: 7413-34-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR