6-Methylchromone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-4H-chromen-4-one

A636353-1G A636353-25G A636353-5G
76806-262EA 46.68 USD
76806-262 76806-264 76806-266
6-Methylchromone 98%
6-Methylchromone

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD00218598
CAS Number: 38445-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR