Diethylene glycol 99%

Supplier: Ambeed
Warning

A635658-100G A635658-25G A635658-500G A635658-1000G
76806-250EA 8 USD
76806-250 76806-252 76806-254 77569-214
Diethylene glycol 99%
Diethylene glycol

Precaución: For research use only.

Formula: C₄H₁₀O₃
MDL Number: MFCD00002882
CAS Number: 111-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR