Dimethoxy(diisobutyl)silane 98%

Supplier: Ambeed
Warning

Synonyms: Diisobutyldimethoxysilane

A634323-100G A634323-25G A634323-5G
76806-236EA 133.68 USD
76806-236 76806-238 76806-240
Dimethoxy(diisobutyl)silane 98%
Dimethoxy(diisobutyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₂₄O₂Si
MW: 204.38 g/mol
MDL Number: MFCD00236510
CAS Number: 17980-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR