Ethyl-4-phenylbutyrate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-phenylbutanoate, 4-Phenylbutyric Acid Ethyl Ester

A629765-1G A629765-5G
76806-216EA 13 USD
76806-216 76806-218
Ethyl-4-phenylbutyrate 98%
Ethyl-4-phenylbutyrate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MDL Number: MFCD00026923
CAS Number: 10031-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR