Di-tert-butyl(methyl)phosphonium tetrafluoroborate 98%

Supplier: Ambeed
Danger

A625506-10G A625506-1G A625506-250MG A625506-25G A625506-5G
76806-200EA 270 USD
76806-200 76806-202 76806-204 76806-206 76806-208
Di-tert-butyl(methyl)phosphonium tetrafluoroborate 98%
Di-tert-butyl(methyl)phosphonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₉H₂₂BF₄P
MDL Number: MFCD03840579
CAS Number: 479094-62-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR