Glipizide 99%

Supplier: Ambeed
Warning

Synonyms: N-[4-(3-Cyclohexylureidosulphonyl)phenethyl]-5-methyl-2-pyrazinecarboxamide, 1-Cyclohexyl-3-{4-[2-(5-methylpyrazine-2-carboxamido)ethyl]phenylsulphonyl}urea, N-(4-(N-(Cyclohexylcarbamoyl)sulphamoyl)phenethyl)-5-methylpyrazine-2-carboxamide

A625260-25MG A625260-1G A625260-1MG A625260-25G A625260-5G
76806-198EA 5.31 USD
76806-198 77602-578 77669-688 77669-690 77669-692
Glipizide 99%
Glipizide

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₅O₄S
MW: 445.54 g/mol
MDL Number: MFCD00072159
CAS Number: 29094-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR