Fmoc-His(3-Bom)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-((benzyloxy)methyl)-1H-imidazol-5-yl)propanoic acid, Fmoc-His(Bom)-OH

A625226-100G A625226-10G A625226-1G A625226-25G A625226-5G
76806-188EA 1438.59 USD
76806-188 76806-190 76806-192 76806-194 76806-196
Fmoc-His(3-Bom)-OH 95%
Fmoc-His(3-Bom)-OH

Precaución: for Research Use Only

Formula: C₂₉H₂₇N₃O₅
MW: 497.551 g/mol
MDL Number: MFCD00077059
CAS Number: 84891-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR