6,7-Dinitro-1,4-dihydroquinoxaline-2,3-dione 98%

Supplier: Ambeed
Danger

Synonyms: DNQX

A622616-25MG A622616-50MG
76806-184EA 71.09 USD
76806-184 76806-186
6,7-Dinitro-1,4-dihydroquinoxaline-2,3-dione 98%
6,7-Dinitro-1,4-dihydroquinoxaline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₄N₄O₆
MW: 252.14 g/mol
MDL Number: MFCD00069257
CAS Number: 2379-57-9
UN: 2926
ADR: 4.1,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR