2-Fluoro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-(pyrrolidine-1-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(pyrolidine-1-carbonyl)phenylboronic acid

A622594-1G A622594-250MG A622594-5G
76806-178EA 168 USD
76806-178 76806-180 76806-182
2-Fluoro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%
2-Fluoro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BFNO₃
MDL Number: MFCD08436030
CAS Number: 874289-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR