Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate

A622302-100MG A622302-25G A622302-5G A622302-250MG
76806-172EA 16.98 USD
76806-172 76806-174 76806-176 77669-684
Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate 97%
Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.10 g/mol
MDL Number: MFCD06659034
CAS Number: 67625-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR