Guanylurea sulfate 98%

Supplier: Ambeed
Warning

Synonyms: Dicyandiamidine Sulphate

A622227-1000G A622227-100G A622227-25G A622227-500G
76806-164EA 154 USD
76806-164 76806-166 76806-168 76806-170
Guanylurea sulfate 98%
Guanylurea sulfate

Precaución: for Research Use Only

Formula: C₂H₈N₄O₅S
MDL Number: MFCD00035565
CAS Number: 591-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR