2,2'-Dipyridylamine 99%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Iminodipyridine, Di(pyridin-2-yl)amine, Chlorpheniramine Related Compound B

A621225-1G A621225-1MG A621225-25G A621225-5G
76806-148EA 25.47 USD
76806-148 76806-150 76806-152 76806-154
2,2'-Dipyridylamine 99%
2,2'-Dipyridylamine

Precaución: for Research Use Only

Formula: C₁₀H₉N₃
MW: 171.2 g/mol
MDL Number: MFCD00006247
CAS Number: 1202-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR