Maltol isobutyrate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-4-oxo-4H-pyran-3-yl isobutyrate, 2-Methyl-4-oxo-4H-pyran-3-yl Isobutyrate, 3-Hydroxy-2-methyl-4-pyrone Isobutyrate, Maltyl isobutyrate

A619097-10G A619097-25G A619097-500G
76806-140EA 9.55 USD
76806-140 76806-142 77669-662
Maltol isobutyrate 95%
Maltol isobutyrate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
MDL Number: MFCD00036606
CAS Number: 65416-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR