Amyl-n-octanoate 98%

Supplier: Ambeed
Warning

Synonyms: n-Octanoic acid Amyl Ester,Pentyl n-Octanoate, Pentyl n-Octanoate, Pentyl octanoate, Amyl n-Octanoate, n-Octanoic Acid Amyl Ester

A618848-100G A618848-25G A618848-500G
76806-134EA 30.77 USD
76806-134 76806-136 76806-138
Amyl-n-octanoate 98%
Amyl-n-octanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
MDL Number: MFCD00048919
CAS Number: 638-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR