2-Methyl-4-quinolinol 98%

Supplier: Ambeed
Warning

A618491-1G A618491-25G A618491-5G
76806-130EA 6 USD
76806-130 76806-132 77880-323
2-Methyl-4-quinolinol 98%
2-Methyl-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD00006758
CAS Number: 607-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR