Ethyl-2-ethoxybenzoate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxybenzoic Acid Ethyl Ester, Ethyl 2-ethoxybenzoate

A618154-100G A618154-25G A618154-5G
76806-124EA 200.52 USD
76806-124 76806-126 76806-128
Ethyl-2-ethoxybenzoate 95%
Ethyl-2-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00142907
CAS Number: 6290-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR