Dipropargylamine 95% (by GC)

Supplier: Ambeed
Danger

Synonyms: Di(2-propynyl)amine

A617612-1G A617612-5G A617612-25G
76806-120EA 25 USD
76806-120 76806-122 77880-319
Dipropargylamine 95% (by GC)
Dipropargylamine

Precaución: for Research Use Only

Formula: C₆H₇N
MW: 93.13 g/mol
MDL Number: MFCD00008574
CAS Number: 6921-28-4
UN: 2734
ADR: 8,3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR