L(+)-Potassium hydrogen tartrate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt

A617434-1000G A617434-100G A617434-25G A617434-500G
76806-112EA 20 USD
76806-112 76806-114 76806-116 76806-118
L(+)-Potassium hydrogen tartrate 98%
L(+)-Potassium hydrogen tartrate

Precaución: for Research Use Only

Formula: KOOC(CHOH)₂COOH
MW: 188.18 g/mol
Melting Pt: 250
MDL Number: MFCD00065392
CAS Number: 868-14-4
Merck Index: 13,07697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR