Dioctylamine 96%

Supplier: Ambeed
Danger

Synonyms: Di-n-octylamine, N-Octyl-1-octanamine

A615019-10G A615019-25G
76806-088EA 11.67 USD
76806-088 77551-844
Dioctylamine 96%
Dioctylamine

Precaución: for Research Use Only

Formula: C₁₆H₃₅N
MW: 241.46 g/mol
MDL Number: MFCD00009557
CAS Number: 1120-48-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR