Diethyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate 98%

Supplier: Ambeed
Warning

A614908-1G A614908-250MG A614908-5G A614908-25G
76806-082EA 44.56 USD
76806-082 76806-084 76806-086 77551-840
Diethyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate 98%
Diethyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate

Precaución: for Research Use Only

Formula: C₈H₁₈NO₅P
MDL Number: MFCD00134233
CAS Number: 124931-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR