Fmoc-Val-Cl 95%

Supplier: Ambeed
Danger

A614270-10G A614270-250MG A614270-25G A614270-5G A614270-1G
76806-074EA 207 USD
76806-074 76806-076 76806-078 76806-080 77880-314
Fmoc-Val-Cl 95%
Fmoc-Val-Cl

Precaución: for Research Use Only

Formula: C₂₀H₂₀NO₃Cl
MDL Number: MFCD00235840
CAS Number: 103321-53-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR