H-D-Leu-Gly-OH 95%

Supplier: Ambeed
Warning

Synonyms: D-Leu-Gly

A614174-1G A614174-5G
76806-070EA 190 USD
76806-070 76806-072
H-D-Leu-Gly-OH 95%
H-D-Leu-Gly-OH

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
MW: 188.22 g/mol
MDL Number: MFCD00067198
CAS Number: 997-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR