Fmoc-Phe-OSu 95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl (((9H-fluoren-9-yl)methoxy)carbonyl)-L-phenylalaninate

A604499-1G A604499-25G A604499-100G A604499-5G
76806-028EA 17.01 USD
76806-028 77669-614 77880-291 77880-292
Fmoc-Phe-OSu 95%
Fmoc-Phe-OSu

Precaución: for Research Use Only

Formula: C₂₈H₂₄N₂O₆
MDL Number: MFCD00062204
CAS Number: 101214-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR