Ethyl Indoline-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl indoline-2-carboxylate, Indoline-2-carboxylic Acid Ethyl Ester

A604305-1G A604305-250MG
76806-024EA 58.35 USD
76806-024 76806-026
Ethyl Indoline-2-carboxylate 95%
Ethyl Indoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
MDL Number: MFCD06797114
CAS Number: 50501-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR