Methyl-2-chloro-6-methoxyisonicotinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-chloro-6-methoxypyridine-4-carboxylate, Methyl 2-chloro-6-methoxyisonicotinate
A603888-1G
A603888-250MG
A603888-25G
A603888-5G
76806-018EA
14
USD
76806-018
76806-020
76806-022
77602-522
Methyl-2-chloro-6-methoxyisonicotinate 98%
Methyl 2-chloro-6-methoxyisonicotinate
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₃ |
MDL Number:
MFCD00173929 CAS Number: 42521-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...