Oxibendazole 98%

Supplier: Ambeed
Warning

A603471-100G A603471-100MG A603471-10MG A603471-1G A603471-250MG A603471-50MG A603471-5G A603471-25G
76805-998EA 695 USD
76805-998 76806-000 76806-002 76806-004 76806-006 76806-008 76806-010 76844-288
Oxibendazole 98%
Oxibendazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₃
MDL Number: MFCD00133728
CAS Number: 20559-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR